Assay Details
| Gene Symbol: Sh3kbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135727.2 | NP_001129199.1 | 5 - 6 | 930 | 64 | |
| NM_021389.6 | NP_067364.2 | 4 - 5 | 495 | 64 | ||
| NR_110969.1 | 4 - 5 | 495 | 64 | |||
| NR_110970.1 | 4 - 5 | 495 | 64 | |||
| GenBank mRNA | AK004636.1 | - | 4 - 5 | 495 | 64 | |
| AK018032.1 | - | 4 - 5 | 511 | 64 | ||
| AK030839.1 | - | 4 - 5 | 496 | 64 | ||
| AK049182.1 | - | 5 - 6 | 931 | 64 | ||
| AK080930.1 | - | 5 - 6 | 795 | 64 | ||
| AK172022.1 | - | 5 - 6 | 911 | 64 | ||
| BC034646.1 | - | 4 - 5 | 499 | 64 | ||
| BC052863.1 | - | 4 - 5 | 444 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sh3kbp1
Genomic Map