Assay Details
| Gene Symbol: Pdzk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146001.1 | NP_001139473.1 | 3 - 4 | 503 | 79 | |
| NM_021517.2 | NP_067492.2 | 3 - 4 | 549 | 79 | ||
| GenBank mRNA | AF220100.1 | - | 3 - 4 | 488 | 79 | |
| AK006269.1 | - | 3 - 4 | 1023 | 79 | ||
| AK012070.1 | - | 3 - 4 | 539 | 79 | ||
| AK014879.1 | - | 3 - 4 | 806 | 79 | ||
| AK029752.1 | - | 3 - 4 | 1007 | 79 | ||
| AK029764.1 | - | 3 - 4 | 1017 | 79 | ||
| AK165415.1 | - | 3 - 4 | 484 | 79 | ||
| BC013512.1 | - | 2 - 3 | 472 | 4234949 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pdzk1
Genomic Map