Assay Details
| Gene Symbol: Psmd14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021526.2 | NP_067501.2 | 5 - 6 | 553 | 75 | |
| GenBank mRNA | AK011974.1 | - | 5 - 6 | 502 | 75 | |
| AK012013.1 | - | 5 - 6 | 484 | 75 | ||
| AK031863.1 | - | 5 - 6 | 491 | 75 | ||
| AK078134.1 | - | 5 - 6 | 505 | 75 | ||
| AK150280.1 | - | 5 - 6 | 489 | 75 | ||
| AK151104.1 | - | 5 - 6 | 536 | 75 | ||
| AK151733.1 | - | 5 - 6 | 536 | 75 | ||
| BC003742.1 | - | 5 - 6 | 458 | 3599671 | 75 | |
| Y13071.1 | - | 5 - 6 | 474 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Psmd14
Genomic Map