Assay Details
| Gene Symbol: Pcbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021567.5 | NP_067542.3 | 1 - 2 | 167 | 63 | |
| GenBank mRNA | AF176328.1 | - | 1 - 2 | 53 | 63 | |
| AK004580.1 | - | 1 - 2 | 118 | 63 | ||
| AK005383.1 | - | 1 - 2 | 116 | 63 | ||
| AK077632.1 | - | 1 - 2 | 117 | 63 | ||
| AK082005.1 | - | 1 - 2 | 117 | 63 | ||
| BC010694.1 | - | 1 - 2 | 56 | 2654291 | 63 | |
| BY259572.1 | - | 1 - 2 | 154 | 63 | ||
| CJ162439.1 | - | 1 - 2 | 164 | 63 | ||
| CJ174803.1 | - | 1 - 2 | 162 | 63 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF176328.1,AK005383.1,AK077632.1,BC010694.1,NM_021567.5 transcripts.
Set Membership: |
|

Pcbp4
Genomic Map