Assay Details
| Gene Symbol: Pcbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021568.2 | NP_067543.2 | 12 - 13 | 1222 | 66 | |
| GenBank mRNA | AF176327.1 | - | 12 - 13 | 1063 | 66 | |
| AK034811.1 | - | 15 - 16 | 1598 | 66 | ||
| AK079564.1 | - | 10 - 11 | 1264 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pcbp3
Genomic Map