Assay Details
| Gene Symbol: Sirt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122765.1 | NP_001116237.1 | 8 - 9 | 680 | 68 | |
| NM_001122766.1 | NP_001116238.1 | 6 - 7 | 517 | 68 | ||
| NM_022432.4 | NP_071877.3 | 9 - 10 | 727 | 68 | ||
| GenBank mRNA | AB221570.1 | - | 9 - 10 | 632 | 68 | |
| AF299337.1 | - | 9 - 10 | 712 | 68 | ||
| AK014042.1 | - | 8 - 9 | 651 | 68 | ||
| AK144431.1 | - | 8 - 9 | 624 | 68 | ||
| AK146412.1 | - | 8 - 9 | 660 | 68 | ||
| AK183534.1 | - | 1 - 2 | 101 | 68 | ||
| BC021439.1 | - | 9 - 10 | 731 | 5039278 | 68 | |
| KF032392.1 | - | 9 - 10 | 669 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sirt2
Genomic Map