Assay Details
Gene Symbol: Scamp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022813.3 | NP_073724.1 | 5 - 6 | 588 | 71 | |
GenBank mRNA | AF295402.1 | - | 5 - 6 | 592 | 71 | |
AK003080.1 | - | 5 - 6 | 590 | 71 | ||
AK161002.1 | - | 5 - 6 | 588 | 71 | ||
AK171766.1 | - | 5 - 6 | 579 | 71 | ||
BC014751.1 | - | 5 - 6 | 565 | 4235556 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
