Assay Details
| Gene Symbol: Slc29a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199113.1 | NP_001186042.1 | 5 - 6 | 556 | 59 | |
| NM_001199114.1 | NP_001186043.1 | 4 - 5 | 395 | 59 | ||
| NM_001199115.1 | NP_001186044.1 | 4 - 5 | 501 | 59 | ||
| NM_001199116.1 | NP_001186045.1 | 4 - 5 | 461 | 59 | ||
| NM_022880.3 | NP_075018.1 | 3 - 4 | 502 | 59 | ||
| GenBank mRNA | AF131212.1 | - | 3 - 4 | 503 | 59 | |
| AK004756.1 | - | 4 - 5 | 447 | 59 | ||
| AK050089.1 | - | 4 - 5 | 486 | 59 | ||
| AK146265.1 | - | 4 - 5 | 396 | 59 | ||
| AK149483.1 | - | 4 - 5 | 417 | 59 | ||
| AK170463.1 | - | 4 - 5 | 433 | 59 | ||
| BC004828.1 | - | 5 - 6 | 519 | 2631728 | 59 | |
| BC006812.1 | - | 3 - 4 | 433 | 3155188 | 59 | |
| CD540408.1 | - | 3 - 4 | 421 | 59 | ||
| CX219686.1 | - | 5 - 6 | 551 | 59 | ||
More Information
Set Membership: |
|

Slc29a1
Genomic Map