Assay Details
| Gene Symbol: Tsc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289575.1 | NP_001276504.1 | 5 - 6 | 590 | 82 | |
| NM_001289576.1 | NP_001276505.1 | 5 - 6 | 590 | 82 | ||
| NM_022887.4 | NP_075025.2 | 5 - 6 | 561 | 82 | ||
| GenBank mRNA | AB047561.1 | - | 3 - 4 | 360 | 82 | |
| AJ271912.1 | - | 5 - 6 | 582 | 82 | ||
| AK030318.1 | - | 5 - 6 | 557 | 82 | ||
| AK078140.1 | - | 5 - 6 | 541 | 82 | ||
| AK083363.1 | - | 5 - 6 | 545 | 82 | ||
| AK147428.1 | - | 5 - 6 | 581 | 82 | ||
| AK154416.1 | - | 5 - 6 | 540 | 82 | ||
| BC052399.1 | - | 4 - 5 | 514 | 5720967 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tsc1
Genomic Map