Assay Details
| Gene Symbol: Tsc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289575.1 | NP_001276504.1 | 6 - 7 | 738 | 61 | |
| NM_001289576.1 | NP_001276505.1 | 6 - 7 | 738 | 61 | ||
| NM_022887.4 | NP_075025.2 | 6 - 7 | 709 | 61 | ||
| GenBank mRNA | AB047561.1 | - | 4 - 5 | 508 | 61 | |
| AJ271912.1 | - | 6 - 7 | 730 | 61 | ||
| AK030318.1 | - | 6 - 7 | 705 | 61 | ||
| AK078140.1 | - | 6 - 7 | 689 | 61 | ||
| AK083363.1 | - | 6 - 7 | 693 | 61 | ||
| AK147428.1 | - | 6 - 7 | 729 | 61 | ||
| AK154416.1 | - | 6 - 7 | 688 | 61 | ||
| BC052399.1 | - | 5 - 6 | 662 | 5720967 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tsc1
Genomic Map