Assay Details
| Gene Symbol: Tsc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289575.1 | NP_001276504.1 | 15 - 16 | 2219 | 69 | |
| NM_001289576.1 | NP_001276505.1 | 15 - 16 | 2216 | 69 | ||
| NM_022887.4 | NP_075025.2 | 15 - 16 | 2187 | 69 | ||
| GenBank mRNA | AB047561.1 | - | 13 - 14 | 1986 | 69 | |
| AJ271912.1 | - | 15 - 16 | 2211 | 69 | ||
| AK030318.1 | - | 15 - 16 | 2184 | 69 | ||
| AK122229.1 | - | 1 - 2 | 526 | 69 | ||
| AK147428.1 | - | 15 - 16 | 2207 | 69 | ||
| BC052399.1 | - | 14 - 15 | 2140 | 5720967 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tsc1
Genomic Map