Assay Details
| Gene Symbol: Pex19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK170175.1 | - | 5 - 6 | 608 | 96 | |
| Y09046.1 | - | 5 - 6 | 600 | 96 | ||
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P118958 | BC012517.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P118958 | BC019767.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P118958 | AK075576.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P118958 | AK170175.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P118958 | AK028449.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P118958 | BC012517.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P118958 | BC019767.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P118958 | AK075576.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P118958 | AK170175.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P118958 | AK028449.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 705020 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 200340 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 569118 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 5102 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 464472 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 336601 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 82574 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 118683 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 110681 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P178712 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P118958 | BC012517.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P118958 | BC019767.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P118958 | AK075576.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P118958 | AK170175.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P118958 | AK028449.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P118958 | BC012517.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P118958 | BC019767.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P118958 | AK075576.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P118958 | AK170175.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P118958 | AK028449.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P427177 | Y09046.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P427177 | BC012517.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P427177 | BC019767.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P427177 | AK075576.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P427177 | AK170175.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20831104 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20831106 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20831117 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20831120 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20831126 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20831136 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20831114 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20831130 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20831128 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20831124 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20831139 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20831140 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20831131 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20831122 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20831118 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20831103 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20831133 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20831129 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20831138 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20831112 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20831134 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20831108 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04355947 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P178712 | - | - |
| Whole Mouse Genome G4122A | A_52_P118958 | BC012517.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P118958 | BC019767.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P118958 | AK075576.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P118958 | AK170175.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P118958 | AK028449.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P118958 | BC012517.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P118958 | BC019767.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P118958 | AK075576.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P118958 | AK170175.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P118958 | AK028449.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P427177 | Y09046.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P427177 | BC012517.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P427177 | BC019767.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P427177 | AK075576.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P427177 | AK170175.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129018 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P178712 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00324807 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00324806 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00756963 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01073767 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01073768 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01073771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20831208 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20831109 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00162588 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20831180 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20831108 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00162589 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20831103 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20831129 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00216296 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20831134 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20831175 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20831106 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20831125 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20831163 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20831132 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20831112 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20831156 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20831159 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00663775 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20831128 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00324807 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20831122 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20831154 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20831160 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20831145 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20831178 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20831142 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20831184 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20831119 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20831139 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20831118 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01109831 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20831177 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20831130 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20831113 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20831105 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00369891 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20831165 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20831110 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00663770 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20831153 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20831188 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20831126 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20831138 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20831182 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01073771 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00907608 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00497794 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20831176 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20831117 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20831120 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01106041 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10351744 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351743 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351739 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351742 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351745 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351740 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351748 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351747 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351746 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351741 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416425_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1442815_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448332_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455208_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10351738 | - | - |
| Mouse Genome 430 2.0 Array | 1442815_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1455208_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448332_at | - | - |
| Mouse Genome 430 2.0 Array | 1448332_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1416425_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1455208_at | - | - |
| Mouse Genome 430 2.0 Array | 1416425_at | - | - |
| Mouse Genome 430 2.0 Array | 1442815_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19298_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19298_at | - | - |
| Mouse Expression Set 430 | 1416425_at | - | - |
| Mouse Expression Set 430 | 1448332_at | - | - |
| Mouse Expression Set 430 | 1416425_PM_at | - | - |
| Mouse Expression Set 430 | 1448332_PM_at | - | - |
| Mouse Expression Set 430 | 1442815_PM_at | - | - |
| Mouse Expression Set 430 | 1455208_PM_at | - | - |
| Mouse Expression Set 430 | 1455208_at | - | - |
| Mouse Expression Set 430 | 1442815_at | - | - |
| Murine Genome U74v2 Set | 95073_PM_at | - | - |
| Murine Genome U74v2 Set | 95074_at | - | - |
| Murine Genome U74v2 Set | 95073_at | - | - |
| Murine Genome U74v2 Set | 95074_PM_at | - | - |
| Murine Genome U74v2 Set | 166423_r_at | - | - |
| Murine Genome U74v2 Set | 166423_PM_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448332_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416425_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416425_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448332_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416425_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448332_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448332_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416425_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455208_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455208_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3140037 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3140037 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003140037 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1239087 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1239087 | - | - |
| mouse-6 v1.1 (Illumina) | scl019298.8_30-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3140037 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl019298.8_30-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3140037 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2970834 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1239087 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1239087 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2970834 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl019298.8_30-S | - | - |

Pex19
Genomic Map