Assay Details
| Gene Symbol: Twsg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023053.3 | NP_075540.1 | 1 - 2 | 121 | 127 | |
| GenBank mRNA | AF292033.1 | - | 60 | 127 | ||
| AJ297390.1 | - | 1 - 2 | 71 | 125 | ||
| AK033504.1 | - | 1 - 2 | 80 | 127 | ||
| AK049646.1 | - | 1 - 2 | 115 | 127 | ||
| AK168547.1 | - | 1 - 2 | 71 | 127 | ||
| AK168633.1 | - | 1 - 2 | 80 | 127 | ||
| BC004850.1 | - | 1 - 2 | 76 | 2810960 | 127 | |
| CJ165542.1 | - | 1 - 2 | 122 | 127 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF292033.1,AJ297390.1,AK033504.1,AK049646.1,AK168547.1,AK168633.1,BC004850.1,NM_023053.3 transcripts.
Set Membership: |
|

Twsg1
Genomic Map