Assay Details
| Gene Symbol: Mrpl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023167.2 | NP_075656.2 | 5 - 6 | 651 | 93 | |
| GenBank mRNA | AB041599.1 | - | 4 - 5 | 93 | ||
| AB049634.1 | - | 5 - 6 | 650 | 93 | ||
| AK002464.1 | - | 5 - 6 | 487 | 93 | ||
| AK003746.2 | - | 5 - 6 | 608 | 93 | ||
| AK216490.1 | - | 3 - 4 | 243 | 93 | ||
| BC037064.1 | - | 5 - 6 | 484 | 93 | ||
| BC039983.1 | - | 5 - 6 | 619 | 4952757 | 93 | |
| BC051685.1 | - | 5 - 6 | 464 | 93 | ||
| BC061095.1 | - | 5 - 6 | 467 | 6588836 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26344984 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26344986 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26344983 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26344985 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26344987 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26344982 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2080396 | BC051685.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2080396 | NM_023167.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2103902 | BC061095.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2103902 | BC023078.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2103902 | BC037064.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2103902 | BC039983.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2103902 | BC051685.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 376898 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 698580 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 45816 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 149303 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P176740 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2080396 | BC051685.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2080396 | NM_023167.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2103902 | BC061095.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2103902 | BC023078.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2103902 | BC037064.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2103902 | BC039983.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2103902 | BC051685.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P213114 | BC061095.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P213114 | BC037064.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P213114 | BC039983.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P213114 | BC051685.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P213114 | AB041599.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344981 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344973 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344992 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344976 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344990 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344984 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344987 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344983 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344986 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344979 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344975 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344985 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344969 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344971 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344988 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P176740 | - | - |
| Whole Mouse Genome G4122A | A_51_P213114 | BC061095.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P213114 | BC037064.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P213114 | BC039983.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P213114 | BC051685.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P213114 | AB041599.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121022 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101547 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P176740 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08049299 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08049309 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08049308 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26344982 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26344986 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26344987 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26344985 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26344984 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26344983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345025 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345000 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345013 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345015 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344975 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08116325 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344999 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344976 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344978 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345007 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344970 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345018 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07955582 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344969 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345012 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345006 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345001 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07868387 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07468339 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344983 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08116326 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344985 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344995 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345003 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344993 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344990 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344996 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345010 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344991 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07731689 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344984 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344986 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08055348 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344971 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344981 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344988 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10583512 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583511 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583513 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583514 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583518 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583509 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583516 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583517 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583515 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583510 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416510_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10583508 | - | - |
| Mouse Genome 430 2.0 Array | 1416510_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1416510_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900001195.mm.1 | NM_023167.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900001195.mm.1 | NM_023167.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900001195.mm.1 | TC0900000183.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900001193.mm.1 | NM_023167.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900001193.mm.1 | NM_023167.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900001193.mm.1 | TC0900000183.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900001197.mm.1 | NM_023167.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900001197.mm.1 | NM_023167.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900001197.mm.1 | TC0900000183.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416510_at | - | - |
| Mouse Expression Set 430 | 1416510_PM_at | - | - |
| Murine Genome U74v2 Set | 95736_at | - | - |
| Murine Genome U74v2 Set | 95736_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416510_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416510_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416510_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416510_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 744893 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2680050 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002680050 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1257408 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1257408 | - | - |
| mouse-6 v1.1 (Illumina) | scl066163.7_0-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2680050 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl066163.7_0-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2680050 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1257408 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1257408 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl066163.7_0-S | - | - |

Mrpl4
Genomic Map