Assay Details
Gene Symbol: Ms4a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023529.2 | NP_076018.2 | 6 - 7 | 665 | 78 | |
GenBank mRNA | AB026046.1 | - | 6 - 7 | 674 | 78 | |
AF237918.1 | - | 6 - 7 | 665 | 78 | ||
AK008019.1 | - | 4 - 5 | 524 | 78 | ||
BC150675.1 | - | 6 - 7 | 662 | 9087586 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
