Assay Details
| Gene Symbol: Txnip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009935.2 | NP_001009935.1 | 6 - 7 | 1273 | 98 | |
| NM_023719.2 | NP_076208.2 | 6 - 7 | 1270 | 98 | ||
| GenBank mRNA | AF173681.1 | - | 6 - 7 | 1037 | 98 | |
| AF282826.1 | - | 6 - 7 | 991 | 98 | ||
| AK004653.2 | - | 6 - 7 | 1270 | 98 | ||
| AK082476.1 | - | 6 - 7 | 1268 | 98 | ||
| AK088870.1 | - | 6 - 7 | 1268 | 98 | ||
| AK089304.1 | - | 6 - 7 | 1273 | 98 | ||
| AK089351.1 | - | 6 - 7 | 1271 | 98 | ||
| AK089403.1 | - | 6 - 7 | 1273 | 98 | ||
| AK155517.1 | - | 6 - 7 | 1271 | 98 | ||
| AK171205.1 | - | 6 - 7 | 1271 | 98 | ||
| AK171312.1 | - | 6 - 7 | 1271 | 98 | ||
| AK171425.1 | - | 6 - 7 | 1271 | 98 | ||
| AK171456.1 | - | 6 - 7 | 1268 | 98 | ||
| AK171464.1 | - | 6 - 7 | 1271 | 98 | ||
| AK171527.1 | - | 6 - 7 | 1271 | 98 | ||
| BC011212.1 | - | 6 - 7 | 1249 | 4193517 | 98 | |
| BC031850.1 | - | 6 - 7 | 1248 | 3591421 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Txnip
Genomic Map