Assay Details
| Gene Symbol: Prl8a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023741.2 | NP_076230.1 | 2 - 3 | 267 | 100 | |
| GenBank mRNA | AF230921.1 | - | 2 - 3 | 253 | 100 | |
| AF230922.1 | - | 2 - 3 | 256 | 100 | ||
| AF230923.1 | - | 3 - 4 | 469 | 100 | ||
| AF466150.1 | - | 2 - 3 | 268 | 100 | ||
| AK005568.1 | - | 2 - 3 | 273 | 100 | ||
| AK167369.1 | - | 2 - 3 | 274 | 100 | ||
| BC100326.1 | - | 2 - 3 | 261 | 30787396 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1953382 | AK005568.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1953382 | AK167369.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1953382 | BC100326.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1953382 | AF230921.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1953382 | AF230922.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 550656 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 447003 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 573503 | AF175305.1 | genbank_mrna_mouse |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P113932 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1953382 | AK005568.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1953382 | AK167369.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1953382 | BC100326.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1953382 | AF230921.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1953382 | AF230922.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902494 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902511 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902478 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902509 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902500 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902489 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902465 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902495 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902504 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902482 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902503 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902487 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902476 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902485 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902467 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902471 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902470 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902505 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902480 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902499 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902475 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902507 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902464 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902468 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902488 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P113932 | - | - |
| Whole Mouse Genome G4122A | A_51_P423637 | AF466150.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P423637 | AK005568.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P423637 | AK167369.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P423637 | BC100326.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P423637 | NM_023741.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115153 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112610 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P113932 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10438320 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10438322 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10438300 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10438314 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10438298 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10438299 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10438312 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10438307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902515 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10451131 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902495 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902522 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902476 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902453 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11004042 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11009560 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902446 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902499 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902505 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902492 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902515 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902428 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10438327 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902473 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902432 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902462 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902451 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902478 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902467 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902517 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902464 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902457 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902511 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902504 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902429 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902503 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902480 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902471 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902494 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10803100 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902441 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902513 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902482 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902444 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902485 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902500 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902487 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902436 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902463 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902434 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902484 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902437 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902456 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902507 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10859824 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902521 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10438322 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902520 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902431 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10561502 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902509 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28902447 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10408392 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408391 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408390 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408389 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408394 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408388 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408393 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424387_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10408386 | - | - |
| Mouse Genome 430 2.0 Array | 1424387_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1424387_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74188_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74188_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300010513.mm.1 | NM_023741.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010513.mm.1 | NM_023741.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010513.mm.1 | TC1300001691.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300010517.mm.1 | NM_023741.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010517.mm.1 | NM_023741.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010517.mm.1 | TC1300001691.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300010514.mm.1 | NM_023741.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010514.mm.1 | NM_023741.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010514.mm.1 | TC1300001691.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424387_PM_at | - | - |
| Mouse Expression Set 430 | 1424387_at | - | - |
| Murine Genome U74v2 Set | 165477_r_at | - | - |
| Murine Genome U74v2 Set | 168863_r_at | - | - |
| Murine Genome U74v2 Set | 165477_PM_r_at | - | - |
| Murine Genome U74v2 Set | 169307_f_at | - | - |
| Murine Genome U74v2 Set | 168863_PM_r_at | - | - |
| Murine Genome U74v2 Set | 169307_PM_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424387_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424387_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424387_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424387_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 917041 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1340088 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1340088 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001340088 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2603927 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2603927 | - | - |
| mouse-6 v1.1 (Illumina) | scl44171.7.1_78-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1340088 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl44171.7.1_78-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1340088 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2786010 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2786010 | - | - |

Prl8a8
Genomic Map