Assay Details
| Gene Symbol: Prl8a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023741.2 | NP_076230.1 | 3 - 4 | 306 | 89 | |
| GenBank mRNA | AF230921.1 | - | 3 - 4 | 292 | 89 | |
| AF230922.1 | - | 3 - 4 | 295 | 89 | ||
| AF230923.1 | - | 4 - 5 | 508 | 89 | ||
| AF466150.1 | - | 3 - 4 | 307 | 89 | ||
| AK005568.1 | - | 3 - 4 | 312 | 89 | ||
| AK167369.1 | - | 3 - 4 | 313 | 89 | ||
| BC100326.1 | - | 3 - 4 | 300 | 30787396 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prl8a8
Genomic Map