Assay Details
| Gene Symbol: Mgea5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023799.3 | NP_076288.1 | 3 - 4 | 510 | 73 | |
| GenBank mRNA | AF132214.1 | - | 3 - 4 | 355 | 73 | |
| AK088774.1 | - | 3 - 4 | 773 | 73 | ||
| AK129188.1 | - | 3 - 4 | 739 | 73 | ||
| AK145227.1 | - | 3 - 4 | 760 | 73 | ||
| BC020093.1 | - | 3 - 4 | 447 | 73 | ||
| BC041109.1 | - | 3 - 4 | 840 | 73 | ||
| BC054821.1 | - | 3 - 4 | 510 | 6515250 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mgea5
Genomic Map