Assay Details
Gene Symbol: Mgea5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023799.3 | NP_076288.1 | 8 - 9 | 1357 | 63 | |
GenBank mRNA | AF132214.1 | - | 8 - 9 | 1202 | 63 | |
AK017997.1 | - | 2 - 3 | 435 | 63 | ||
AK088774.1 | - | 8 - 9 | 1620 | 63 | ||
AK129188.1 | - | 8 - 9 | 1586 | 63 | ||
AK145227.1 | - | 8 - 9 | 1607 | 63 | ||
BC020093.1 | - | 8 - 9 | 1294 | 63 | ||
BC041109.1 | - | 8 - 9 | 1687 | 63 | ||
BC054821.1 | - | 8 - 9 | 1357 | 6515250 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
