Assay Details
| Gene Symbol: Slco3a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038643.1 | NP_001033732.1 | 4 - 5 | 1201 | 64 | |
| NM_023908.2 | NP_076397.2 | 4 - 5 | 1201 | 64 | ||
| GenBank mRNA | AF226324.1 | - | 4 - 5 | 1099 | 64 | |
| AK031280.1 | - | 4 - 5 | 1201 | 64 | ||
| AK037938.1 | - | 4 - 5 | 1016 | 64 | ||
| AK046945.1 | - | 3 - 4 | 561 | 64 | ||
| AK151781.1 | - | 4 - 5 | 1171 | 64 | ||
| AK152488.1 | - | 4 - 5 | 1035 | 64 | ||
| AK166266.1 | - | 4 - 5 | 972 | 64 | ||
| BC025059.1 | - | 4 - 5 | 1034 | 2598913 | 64 | |
| BC094464.1 | - | 4 - 5 | 993 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slco3a1
Genomic Map