Assay Details
| Gene Symbol: Scyl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023912.2 | NP_076401.1 | 5 - 6 | 922 | 93 | |
| GenBank mRNA | AF276514.1 | - | 5 - 6 | 922 | 93 | |
| AK019240.1 | - | 5 - 6 | 915 | 93 | ||
| AK134356.1 | - | 5 - 6 | 921 | 93 | ||
| AK144134.1 | - | 5 - 6 | 908 | 93 | ||
| AK145869.1 | - | 5 - 6 | 942 | 93 | ||
| AK161813.1 | - | 5 - 6 | 921 | 93 | ||
| BC034519.1 | - | 5 - 6 | 908 | 3672524 | 93 | |
| EF437375.1 | - | 1 - 2 | 101 | 93 | ||
| HM856325.1 | - | 5 - 6 | 922 | 93 | ||
| HM856326.1 | - | 5 - 6 | 922 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Scyl1
Genomic Map