Assay Details
| Gene Symbol: Scyl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023912.2 | NP_076401.1 | 10 - 11 | 1650 | 71 | |
| GenBank mRNA | AF276514.1 | - | 10 - 11 | 1650 | 71 | |
| AK019240.1 | - | 10 - 11 | 1643 | 71 | ||
| AK134356.1 | - | 10 - 11 | 1649 | 71 | ||
| AK144134.1 | - | 10 - 11 | 1636 | 71 | ||
| AK145869.1 | - | 10 - 11 | 1670 | 71 | ||
| AK161813.1 | - | 10 - 11 | 1649 | 71 | ||
| BC034519.1 | - | 10 - 11 | 1636 | 3672524 | 71 | |
| HM856325.1 | - | 10 - 11 | 1650 | 71 | ||
| HM856326.1 | - | 10 - 11 | 1650 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Scyl1
Genomic Map