Assay Details
| Gene Symbol: Riok3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024182.4 | NP_077144.2 | 1 - 2 | 128 | 110 | |
| GenBank mRNA | AK004748.1 | - | 1 - 2 | 220 | 110 | |
| AK131635.1 | - | 1 - 2 | 132 | 110 | ||
| AK134327.1 | - | 1 - 2 | 220 | 110 | ||
| AK147074.1 | - | 1 - 2 | 129 | 110 | ||
| AK167309.1 | - | 1 - 2 | 224 | 110 | ||
| AK170882.1 | - | 1 - 2 | 208 | 110 | ||
| BC033271.1 | - | 1 - 2 | 166 | 4016197 | 110 | |
More Information
Updated Mapping Information
Set Membership: |
|

Riok3
Genomic Map