Assay Details
| Gene Symbol: Riok3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024182.4 | NP_077144.2 | 2 - 3 | 241 | 112 | |
| GenBank mRNA | AK004748.1 | - | 2 - 3 | 333 | 112 | |
| AK131635.1 | - | 2 - 3 | 245 | 112 | ||
| AK134327.1 | - | 2 - 3 | 333 | 112 | ||
| AK147074.1 | - | 2 - 3 | 242 | 112 | ||
| AK167309.1 | - | 2 - 3 | 337 | 112 | ||
| AK170882.1 | - | 2 - 3 | 321 | 112 | ||
| BC033271.1 | - | 2 - 3 | 279 | 4016197 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|

Riok3
Genomic Map