Assay Details
| Gene Symbol: Slc25a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024211.3 | NP_077173.1 | 2 - 3 | 420 | 64 | |
| GenBank mRNA | AI931249.1 | - | 2 - 3 | 381 | 64 | |
| AK003957.1 | - | 1 - 2 | 59 | 64 | ||
| AK009487.1 | - | 2 - 3 | 366 | 64 | ||
| AK009824.1 | - | 2 - 3 | 320 | 64 | ||
| AK168951.1 | - | 2 - 3 | 345 | 64 | ||
| AK181540.1 | - | 1 - 2 | 102 | 64 | ||
| AK187524.1 | - | 1 - 2 | 102 | 64 | ||
| AK202646.1 | - | 1 - 2 | 102 | 64 | ||
| AK212146.1 | - | 1 - 2 | 92 | 64 | ||
| AK213879.1 | - | 1 - 2 | 102 | 64 | ||
| AK215545.1 | - | 1 - 2 | 92 | 64 | ||
| BC003455.1 | - | 2 - 3 | 396 | 2648336 | 64 | |
| BC019631.1 | - | 2 - 3 | 374 | 4023179 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27901827 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27901831 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27901828 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27901832 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27901829 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267454 | BC019631.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267454 | AK206341.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267454 | AK212009.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267454 | AK009487.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267454 | NM_024211.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267454 | BC019631.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267454 | AK206341.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267454 | AK212009.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267454 | AK009487.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267454 | NM_024211.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 449718 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267454 | BC019631.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267454 | AK206341.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267454 | AK212009.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267454 | AK009487.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267454 | NM_024211.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267454 | BC019631.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267454 | AK206341.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267454 | AK212009.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267454 | AK009487.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267454 | NM_024211.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P267454 | BC019631.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P267454 | AK206341.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P267454 | AK212009.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P267454 | AK009487.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P267454 | NM_024211.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P267454 | BC019631.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P267454 | AK206341.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P267454 | AK212009.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P267454 | AK009487.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P267454 | NM_024211.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901806 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901819 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901823 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901828 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901830 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901801 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901807 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901800 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901824 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901816 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901810 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901832 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901808 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901799 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901833 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901821 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901813 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901818 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901826 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901804 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901822 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901812 | - | - |
| Whole Mouse Genome G4122A | A_51_P267454 | BC019631.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267454 | AK206341.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267454 | AK212009.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267454 | AK009487.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267454 | NM_024211.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267454 | BC019631.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267454 | AK206341.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267454 | AK212009.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267454 | AK009487.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267454 | NM_024211.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110085 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09578942 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09578931 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09578912 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09578929 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27901828 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27901827 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27901832 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27901829 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27901831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901768 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09578916 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09578950 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09578914 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901824 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901823 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09578910 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901801 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09578946 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901800 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901813 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901830 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901827 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09258669 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901799 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901810 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09578929 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901818 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901808 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901822 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09578956 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10387924 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387929 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387925 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387923 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387930 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387926 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387931 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387927 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387928 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426586_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10387922 | - | - |
| Mouse Genome 430 2.0 Array | 1426586_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1426586_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67863_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67863_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100028672.mm.1 | NM_024211.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100028672.mm.1 | NM_024211.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100028672.mm.1 | TC1100003106.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100028675.mm.1 | NM_024211.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100028675.mm.1 | NM_024211.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100028675.mm.1 | TC1100003106.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1426586_PM_at | - | - |
| Mouse Expression Set 430 | 1426586_at | - | - |
| Murine Genome U74v2 Set | 160295_at | - | - |
| Murine Genome U74v2 Set | 160295_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426586_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426586_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426586_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426586_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 325739 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 130097 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 130097 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 770685 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000770685 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000130097 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2600053 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1236772 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1236772 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2600053 | - | - |
| mouse-6 v1.1 (Illumina) | scl0001455.1_38-S | - | - |
| mouse-6 v1.1 (Illumina) | scl39988.9.1_13-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 130097 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 770685 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0001455.1_38-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl39988.9.1_13-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 130097 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 770685 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3010110 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3010109 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2600053 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3010109 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3010110 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2600053 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl39988.9.1_13-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0001455.1_38-S | - | - |

Slc25a11
Genomic Map