Assay Details
| Gene Symbol: Slc25a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024211.3 | NP_077173.1 | 4 - 5 | 724 | 68 | |
| GenBank mRNA | AK003957.1 | - | 3 - 4 | 363 | 68 | |
| AK009487.1 | - | 4 - 5 | 670 | 68 | ||
| AK009824.1 | - | 4 - 5 | 624 | 68 | ||
| AK168951.1 | - | 4 - 5 | 649 | 68 | ||
| BC003455.1 | - | 4 - 5 | 700 | 2648336 | 68 | |
| BC019631.1 | - | 4 - 5 | 678 | 4023179 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc25a11
Genomic Map