Assay Details
| Gene Symbol: Mrps15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025544.2 | NP_079820.2 | 3 - 4 | 399 | 76 | |
| GenBank mRNA | AB049947.1 | - | 3 - 4 | 275 | 76 | |
| AK003126.1 | - | 3 - 4 | 320 | 76 | ||
| AK005131.2 | - | 3 - 4 | 350 | 76 | ||
| AK008623.1 | - | 3 - 4 | 351 | 76 | ||
| AK010330.1 | - | 3 - 4 | 362 | 76 | ||
| AK050396.1 | - | 3 - 4 | 403 | 76 | ||
| AK140877.1 | - | 3 - 4 | 404 | 76 | ||
| BC029193.1 | - | 3 - 4 | 318 | 4457237 | 76 | |
| BC055861.1 | - | 3 - 4 | 298 | 3587953 | 76 | |
| CF582282.1 | - | 5 - 6 | 76 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Mrps15
Genomic Map