Assay Details
| Gene Symbol: Acot11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025590.4 | NP_079866.2 | 1 - 2 | 144 | 77 | |
| GenBank mRNA | AF416923.1 | - | 1 - 2 | 82 | 77 | |
| AK031588.1 | - | 1 - 2 | 61 | 77 | ||
| AK048670.1 | - | 1 - 2 | 144 | 77 | ||
| AK052332.1 | - | 1 - 2 | 160 | 77 | ||
| BC042492.1 | - | 1 - 2 | 79 | 4240966 | 77 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF416923.1,AK031588.1,AK048670.1,AK052332.1,BC042492.1,NM_025590.4 transcripts.
Set Membership: |
|

Acot11
Genomic Map