Assay Details
| Gene Symbol: Acot11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025590.4 | NP_079866.2 | 2 - 3 | 363 | 93 | |
| GenBank mRNA | AF416923.1 | - | 2 - 3 | 301 | 93 | |
| AK031588.1 | - | 2 - 3 | 280 | 93 | ||
| AK038546.1 | - | 2 - 3 | 301 | 93 | ||
| AK048670.1 | - | 2 - 3 | 363 | 93 | ||
| AK052332.1 | - | 2 - 3 | 379 | 93 | ||
| AK122350.1 | - | 1 - 2 | 150 | 93 | ||
| BC042492.1 | - | 2 - 3 | 298 | 4240966 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acot11
Genomic Map