Assay Details
| Gene Symbol: Acot11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025590.4 | NP_079866.2 | 11 - 12 | 1270 | 79 | |
| GenBank mRNA | AF416923.1 | - | 11 - 12 | 1208 | 79 | |
| AK031588.1 | - | 11 - 12 | 1187 | 79 | ||
| AK038546.1 | - | 11 - 12 | 1208 | 79 | ||
| AK048670.1 | - | 11 - 12 | 1270 | 79 | ||
| AK052332.1 | - | 11 - 12 | 1286 | 79 | ||
| AK122350.1 | - | 10 - 11 | 1057 | 79 | ||
| AK170189.1 | - | 4 - 5 | 293 | 79 | ||
| AK171255.1 | - | 4 - 5 | 294 | 79 | ||
| BC042492.1 | - | 11 - 12 | 1205 | 4240966 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acot11
Genomic Map