Assay Details
| Gene Symbol: Mto1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026658.2 | NP_080934.2 | 2 - 3 | 502 | 84 | |
| GenBank mRNA | AF369902.1 | - | 2 - 3 | 492 | 84 | |
| AK019088.1 | - | 2 - 3 | 502 | 84 | ||
| AK202743.1 | - | 1 - 2 | 113 | 84 | ||
| BC063256.1 | - | 2 - 3 | 488 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mto1
Genomic Map