Assay Details
| Gene Symbol: Mrpl44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081210.1 | NP_001074679.1 | 2 - 3 | 758 | 70 | |
| GenBank mRNA | AK007652.1 | - | 1 - 2 | 511 | 70 | |
| AK146024.1 | - | 2 - 3 | 570 | 70 | ||
| AK150537.1 | - | 2 - 3 | 520 | 70 | ||
| AK151854.1 | - | 2 - 3 | 520 | 70 | ||
| AK156814.1 | - | 2 - 3 | 673 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P198009 | AK146024.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P198009 | AK150537.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P198009 | AK151854.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P198009 | AK156814.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P198009 | BC019727.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P198009 | AK146024.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P198009 | AK150537.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P198009 | AK151854.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P198009 | AK156814.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P198009 | BC019727.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 446476 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 155111 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 675222 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 174858 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P160833 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P132616 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P198009 | AK146024.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P198009 | AK150537.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P198009 | AK151854.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P198009 | AK156814.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P198009 | BC019727.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P198009 | AK146024.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P198009 | AK150537.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P198009 | AK151854.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P198009 | AK156814.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P198009 | BC019727.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P160833 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P132616 | - | - |
| Whole Mouse Genome G4122A | A_52_P198009 | AK146024.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P198009 | AK150537.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P198009 | AK151854.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P198009 | AK156814.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P198009 | BC019727.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P198009 | AK146024.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P198009 | AK150537.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P198009 | AK151854.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P198009 | AK156814.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P198009 | BC019727.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P668030 | AK019986.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P668030 | AK146024.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P668030 | AK150537.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P668030 | AK151854.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P668030 | AK156814.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P471940 | AK146024.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P471940 | AK150537.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P471940 | AK151854.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P471940 | AK156814.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P471940 | NM_001081210.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P160833 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P132616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20377661 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377622 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377637 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00638482 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377653 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377666 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377629 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377673 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377624 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377690 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377681 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377661 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377688 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00112384 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377657 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377640 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377659 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377627 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377711 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00112391 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377634 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377633 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377686 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377677 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377626 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377672 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377664 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377685 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377623 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377674 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377671 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00180969 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377667 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377643 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377669 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00054887 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377632 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377663 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00892324 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377625 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377648 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377687 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00881429 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377701 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377695 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377650 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377692 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377679 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377628 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377705 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377631 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377647 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20377699 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10347777 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10347778 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10347776 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10347775 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426651_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1437391_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430990_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452144_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10347774 | - | - |
| Mouse Genome 430 2.0 Array | 1430990_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1430990_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1437391_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1452144_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1426651_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1437391_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1426651_at | - | - |
| Mouse Genome 430 2.0 Array | 1452144_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69163_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69163_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100005900.mm.1 | NM_001081210.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100005900.mm.1 | NM_001081210.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100005900.mm.1 | TC0100000688.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100005901.mm.1 | NM_001081210.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100005901.mm.1 | NM_001081210.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100005901.mm.1 | TC0100000688.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1452144_PM_a_at | - | - |
| Mouse Expression Set 430 | 1426651_at | - | - |
| Mouse Expression Set 430 | 1426651_PM_at | - | - |
| Mouse Expression Set 430 | 1452144_a_at | - | - |
| Mouse Expression Set 430 | 1437391_PM_x_at | - | - |
| Mouse Expression Set 430 | 1437391_x_at | - | - |
| Murine Genome U74v2 Set | 97296_PM_at | - | - |
| Murine Genome U74v2 Set | 97296_at | - | - |
| Mouse Genome 430A 2.0 Array | 1437391_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1437391_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452144_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452144_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426651_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426651_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1437391_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452144_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452144_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426651_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1437391_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426651_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430990_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430990_PM_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 700442 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1227033 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1227033 | - | - |

Mrpl44
Genomic Map