Assay Details
| Gene Symbol: Usp15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301628.1 | NP_001288557.1 | 5 - 6 | 723 | 70 | |
| NM_027604.4 | NP_081880.2 | 5 - 6 | 723 | 70 | ||
| GenBank mRNA | AF468037.1 | - | 5 - 6 | 627 | 70 | |
| AK046332.1 | - | 5 - 6 | 634 | 70 | ||
| AK052398.1 | - | 4 - 5 | 539 | 70 | ||
| AK122301.1 | - | 5 - 6 | 638 | 70 | ||
| AK133852.1 | - | 5 - 6 | 632 | 70 | ||
| AK145749.1 | - | 5 - 6 | 627 | 70 | ||
| AK161469.1 | - | 5 - 6 | 666 | 70 | ||
| AK161957.1 | - | 5 - 6 | 671 | 70 | ||
| AK163315.1 | - | 5 - 6 | 652 | 70 | ||
| AK164382.1 | - | 5 - 6 | 646 | 70 | ||
| AK168755.1 | - | 5 - 6 | 658 | 70 | ||
| BC050042.1 | - | 5 - 6 | 639 | 30022307 | 70 | |
More Information
Set Membership: |
|

Usp15
Genomic Map