Assay Details
| Gene Symbol: Scpep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029023.3 | NP_083299.3 | 2 - 3 | 297 | 62 | |
| GenBank mRNA | AF330052.1 | - | 2 - 3 | 253 | 62 | |
| AK014680.1 | - | 2 - 3 | 298 | 62 | ||
| AK050181.1 | - | 2 - 3 | 235 | 62 | ||
| AK153225.1 | - | 2 - 3 | 285 | 62 | ||
| AK153485.1 | - | 2 - 3 | 242 | 62 | ||
| BC004847.1 | - | 2 - 3 | 287 | 2650587 | 62 | |
| BC021399.1 | - | 2 - 3 | 252 | 4193674 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28003084 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28003082 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28003085 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990210 | BC021399.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990210 | BC004847.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P136714 | BC021399.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P136714 | BC004847.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 815048 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 229463 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 861826 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 965534 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 791078 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 14295 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 650950 | NM_001011737.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 268604 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 882375 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 566330 | AK033940.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 945641 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 297390 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 785073 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 135267 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 629270 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 461439 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 249574 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 137324 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 550681 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 58570 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 79641 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 110963 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 19244 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P174119 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P121589 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990210 | BC021399.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990210 | BC004847.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P118415 | BC021399.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P118415 | BC004847.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P118417 | BC021399.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P118417 | BC004847.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28003081 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28003093 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28003086 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28003103 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28003091 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28003089 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28003078 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28003084 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28003076 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P121589 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07435026 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02389702 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P174119 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02389743 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02389710 | - | - |
| Whole Mouse Genome G4122A | A_51_P118417 | BC021399.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P118417 | BC004847.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P118415 | BC021399.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P118415 | BC004847.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136714 | BC021399.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136714 | BC004847.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P174119 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P121589 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P02389743 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09393055 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09393060 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09393056 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28003084 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28003085 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28003082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28002911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28002961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28002984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28002930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28002926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28002957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28002946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28002969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28002935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28002979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28002992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28002959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28002971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28002953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28002998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28002987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28002923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28002976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28002963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28002918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28002929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28002967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28002913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28002986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28002949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28002940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28002920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28002931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28002941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28002942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28002999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28002944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28002933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28002997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28002951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28002996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28003067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28002954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28002955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28002981 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28003071 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28003046 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28003073 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28003064 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09490294 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09393055 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28003067 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09547370 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28003051 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28003081 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09069690 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28003062 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28003058 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28003069 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09490293 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09421262 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28003075 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28003066 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28003065 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09526716 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28003015 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28003057 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28003079 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28003053 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09445389 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09335392 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28003047 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28003103 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28003086 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28003005 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28003007 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09335394 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28003014 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28003092 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09063995 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28003076 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28003089 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28003094 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28003091 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10389725 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389726 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389728 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389721 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389730 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389722 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389723 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389729 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389720 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389727 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389731 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389724 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455908_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426555_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10389719 | - | - |
| Mouse Genome 430 2.0 Array | 1455908_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1426555_at | - | - |
| Mouse Genome 430 2.0 Array | 1455908_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1426555_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74617_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74617_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100032105.mm.1 | NM_029023.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100032105.mm.1 | NM_029023.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100032105.mm.1 | TC1100003506.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100032106.mm.1 | NM_029023.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100032106.mm.1 | NM_029023.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100032106.mm.1 | TC1100003506.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100032104.mm.1 | NM_029023.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100032104.mm.1 | NM_029023.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100032104.mm.1 | TC1100003506.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100032107.mm.1 | NM_029023.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100032107.mm.1 | NM_029023.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100032107.mm.1 | TC1100003506.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1426555_PM_at | - | - |
| Mouse Expression Set 430 | 1455908_PM_a_at | - | - |
| Mouse Expression Set 430 | 1426555_at | - | - |
| Mouse Expression Set 430 | 1455908_a_at | - | - |
| Murine Genome U74v2 Set | 98042_PM_at | - | - |
| Murine Genome U74v2 Set | 98042_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426555_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426555_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455908_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455908_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455908_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455908_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426555_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426555_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 509024 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3610576 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003610576 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2736968 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2736968 | - | - |
| mouse-6 v1.1 (Illumina) | scl39738.13.1_56-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3610576 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl39738.13.1_56-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3610576 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2736968 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2736968 | - | - |

Scpep1
Genomic Map