Assay Details
| Gene Symbol: Mrps5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029963.2 | NP_084239.1 | 5 - 6 | 644 | 144 | |
| GenBank mRNA | AB049941.1 | - | 5 - 6 | 644 | 144 | |
| AK083366.1 | - | 5 - 6 | 646 | 144 | ||
| AK155101.1 | - | 5 - 6 | 660 | 144 | ||
| BC021947.1 | - | 5 - 6 | 642 | 5098933 | 144 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mrps5
Genomic Map