Assay Details
| Gene Symbol: Upf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122829.1 | NP_001116301.1 | 14 - 15 | 2264 | 91 | |
| NM_030680.2 | NP_109605.2 | 14 - 15 | 2231 | 91 | ||
| GenBank mRNA | AF182947.1 | - | 15 - 16 | 2220 | 91 | |
| AF322655.1 | - | 14 - 15 | 1951 | 91 | ||
| AK052517.1 | - | 3 - 4 | 336 | 91 | ||
| AK148196.1 | - | 15 - 16 | 2241 | 91 | ||
| BC030916.1 | - | 2 - 3 | 242 | 91 | ||
| BC052149.1 | - | 14 - 15 | 2169 | 6332288 | 91 | |
| BC056442.1 | - | 14 - 15 | 2264 | 5702493 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Upf1
Genomic Map