Assay Details
| Gene Symbol: Trim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271725.1 | NP_001258654.1 | 3 - 4 | 570 | 63 | |
| NM_001271726.1 | NP_001258655.1 | 3 - 4 | 606 | 63 | ||
| NM_001271727.1 | NP_001258656.1 | 3 - 4 | 875 | 63 | ||
| NM_001271728.1 | NP_001258657.1 | 3 - 4 | 673 | 63 | ||
| NM_030706.3 | NP_109631.2 | 3 - 4 | 669 | 63 | ||
| GenBank mRNA | AB043550.1 | - | 3 - 4 | 653 | 63 | |
| AF220017.1 | - | 3 - 4 | 518 | 63 | ||
| AK039741.1 | - | 3 - 4 | 667 | 63 | ||
| AK045410.1 | - | 3 - 4 | 666 | 63 | ||
| AK079415.1 | - | 3 - 4 | 820 | 63 | ||
| AK141419.1 | - | 3 - 4 | 516 | 63 | ||
| AK147275.1 | - | 3 - 4 | 783 | 63 | ||
| AK147395.1 | - | 3 - 4 | 664 | 63 | ||
| AK164350.1 | - | 2 - 3 | 372 | 63 | ||
| AK220343.1 | - | 3 - 4 | 874 | 63 | ||
| BC058668.1 | - | 3 - 4 | 637 | 63 | ||
| BC058961.1 | - | 3 - 4 | 673 | 6826143 | 63 | |
| CD802800.1 | - | 3 - 4 | 532 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trim2
Genomic Map