Assay Details
| Gene Symbol: Trim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271725.1 | NP_001258654.1 | 5 - 6 | 903 | 76 | |
| NM_001271726.1 | NP_001258655.1 | 5 - 6 | 939 | 76 | ||
| NM_001271727.1 | NP_001258656.1 | 5 - 6 | 1208 | 76 | ||
| NM_001271728.1 | NP_001258657.1 | 5 - 6 | 1006 | 76 | ||
| NM_030706.3 | NP_109631.2 | 5 - 6 | 1002 | 76 | ||
| GenBank mRNA | AB043550.1 | - | 5 - 6 | 986 | 76 | |
| AF220017.1 | - | 5 - 6 | 851 | 76 | ||
| AK039741.1 | - | 5 - 6 | 1000 | 76 | ||
| AK045410.1 | - | 5 - 6 | 999 | 76 | ||
| AK079415.1 | - | 5 - 6 | 1153 | 76 | ||
| AK141419.1 | - | 5 - 6 | 849 | 76 | ||
| AK147275.1 | - | 5 - 6 | 1116 | 76 | ||
| AK147395.1 | - | 5 - 6 | 997 | 76 | ||
| AK164350.1 | - | 4 - 5 | 76 | |||
| AK220343.1 | - | 5 - 6 | 1207 | 76 | ||
| BC058961.1 | - | 5 - 6 | 1006 | 6826143 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trim2
Genomic Map