Assay Details
| Gene Symbol: Polh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030715.3 | NP_109640.1 | 8 - 9 | 1241 | 64 | |
| GenBank mRNA | AB027128.1 | - | 8 - 9 | 1267 | 64 | |
| AK036296.1 | - | 8 - 9 | 1242 | 64 | ||
| BC049159.1 | - | 8 - 9 | 1211 | 64 | ||
| BC128366.1 | - | 7 - 8 | 1023 | 40111620 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Polh
Genomic Map