Assay Details
| Gene Symbol: Pacsin3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289678.1 | NP_001276607.1 | 2 - 3 | 179 | 61 | |
| NM_028733.4 | NP_083009.1 | 2 - 3 | 309 | 61 | ||
| NM_030880.3 | NP_112019.2 | 2 - 3 | 175 | 61 | ||
| GenBank mRNA | AF149824.1 | - | 2 - 3 | 93 | 61 | |
| AF242531.1 | - | 1 - 2 | 78 | 61 | ||
| AK049166.1 | - | 2 - 3 | 310 | 61 | ||
| AK141063.1 | - | 2 - 3 | 152 | 61 | ||
| BC003884.1 | - | 2 - 3 | 116 | 3497435 | 61 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF149824.1,AF242531.1,AK049166.1,AK141063.1,BC003884.1,NM_001289678.1,NM_028733.4,NM_030880.3 transcripts.
Set Membership: |
|

Pacsin3
Genomic Map