Assay Details
| Gene Symbol: Ogfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031373.3 | NP_113550.3 | 2 - 3 | 280 | 71 | |
| GenBank mRNA | AF303894.1 | - | 2 - 3 | 228 | 71 | |
| AK008230.1 | - | 2 - 3 | 245 | 71 | ||
| AK149146.1 | - | 2 - 3 | 227 | 71 | ||
| AK150115.1 | - | 2 - 3 | 213 | 71 | ||
| BC003875.1 | - | 2 - 3 | 211 | 3495186 | 71 | |
| BC049965.1 | - | 2 - 3 | 226 | 3155091 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ogfr
Genomic Map