Assay Details
| Gene Symbol: Ogfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031373.3 | NP_113550.3 | 4 - 5 | 442 | 101 | |
| GenBank mRNA | AF303894.1 | - | 4 - 5 | 390 | 101 | |
| AK008230.1 | - | 4 - 5 | 407 | 101 | ||
| AK149146.1 | - | 4 - 5 | 389 | 101 | ||
| AK150115.1 | - | 4 - 5 | 375 | 101 | ||
| BC003875.1 | - | 4 - 5 | 373 | 3495186 | 101 | |
| BC049965.1 | - | 4 - 5 | 388 | 3155091 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ogfr
Genomic Map