Assay Details
| Gene Symbol: Strn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172098.1 | NP_001165569.1 | 3 - 4 | 671 | 111 | |
| NM_052973.2 | NP_443205.1 | 3 - 4 | 671 | 111 | ||
| GenBank mRNA | AF307777.1 | - | 3 - 4 | 655 | 111 | |
| AK139183.1 | - | 3 - 4 | 655 | 111 | ||
| AK164967.1 | - | 3 - 4 | 673 | 111 | ||
| AK165908.1 | - | 3 - 4 | 355 | 111 | ||
| BC138055.1 | - | 3 - 4 | 622 | 8861074 | 111 | |
| BC145127.1 | - | 3 - 4 | 622 | 9053675 | 111 | |
| EF685152.1 | - | 3 - 4 | 655 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Strn3
Genomic Map