Assay Details
| Gene Symbol: Slc15a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053079.2 | NP_444309.2 | 8 - 9 | 670 | 59 | |
| GenBank mRNA | AF205540.1 | - | 8 - 9 | 669 | 59 | |
| AK085471.1 | - | 6 - 7 | 1777 | 59 | ||
| AK136428.1 | - | 8 - 9 | 671 | 59 | ||
| BC116248.1 | - | 8 - 9 | 660 | 40093541 | 59 | |
| BC116249.1 | - | 8 - 9 | 660 | 40093542 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc15a1
Genomic Map