Assay Details
| Gene Symbol: Erc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053204.2 | NP_444434.2 | 7 - 8 | 1976 | 105 | |
| NR_104484.1 | 7 - 8 | 1976 | 105 | |||
| GenBank mRNA | AF340028.1 | - | 7 - 8 | 1960 | 105 | |
| AF340029.1 | - | 7 - 8 | 1960 | 105 | ||
| AY316692.1 | - | 5 - 6 | 1408 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Erc1
Genomic Map