Assay Details
| Gene Symbol: Erc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053204.2 | NP_444434.2 | 16 - 17 | 3199 | 61 | |
| GenBank mRNA | AF340029.1 | - | 16 - 17 | 3183 | 61 | |
| AY316692.1 | - | 14 - 15 | 2631 | 61 | ||
More Information
Set Membership: |
|

Erc1
Genomic Map