Assay Details
| Gene Symbol: Erc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053204.2 | NP_444434.2 | 19 - 20 | 3606 | 114 | |
| GenBank mRNA | AF340029.1 | - | 19 - 20 | 3590 | 114 | |
| AY316692.1 | - | 17 - 18 | 3038 | 114 | ||
More Information
Set Membership: |
|

Erc1
Genomic Map