Assay Details
| Gene Symbol: Mvp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080638.3 | NP_542369.2 | 11 - 12 | 2118 | 77 | |
| GenBank mRNA | AF210456.1 | - | 11 - 12 | 2120 | 77 | |
| AK019080.1 | - | 1 - 2 | 313 | 77 | ||
| AK088046.1 | - | 11 - 12 | 2103 | 77 | ||
| AK167899.1 | - | 11 - 12 | 2094 | 77 | ||
| AK168100.1 | - | 11 - 12 | 2094 | 77 | ||
| AK182246.1 | - | 1 - 2 | 145 | 77 | ||
| BC006709.1 | - | 11 - 12 | 2083 | 3584439 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mvp
Genomic Map