Assay Details
| Gene Symbol: Mtmr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133215.1 | NP_573478.1 | 7 - 8 | 622 | 74 | |
| GenBank mRNA | AF262986.1 | - | 7 - 8 | 622 | 74 | |
| AK146641.1 | - | 7 - 8 | 639 | 74 | ||
| AK217568.1 | - | 1 - 2 | 68 | 74 | ||
| BC058091.1 | - | 7 - 8 | 620 | 6822365 | 74 | |
| BC058364.1 | - | 7 - 8 | 620 | 6820754 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mtmr4
Genomic Map